Assay Details
| Gene Symbol: LOC4339037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784233.2 | XP_015639719.1 | 14 - 15 | 4444 | 115 | |
| XM_015784234.2 | XP_015639720.1 | 14 - 15 | 4441 | 115 | ||
| GenBank mRNA | AK064382.1 | - | 5 - 6 | 1350 | 115 | |
More Information
Set Membership: |
|

LOC4339037
Genomic Map