Assay Details
| Gene Symbol: LOC4339037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784234.2 | XP_015639720.1 | 15 - 16 | 4576 | 130 | |
| GenBank mRNA | AK064382.1 | - | 6 - 7 | 1485 | 130 | |
More Information
Set Membership: |
|

LOC4339037
Genomic Map