Assay Details
| Gene Symbol: LOC4339037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784233.2 | XP_015639719.1 | 16 - 17 | 4656 | 113 | |
| XM_015784234.2 | XP_015639720.1 | 16 - 17 | 4650 | 113 | ||
| GenBank mRNA | AK064382.1 | - | 7 - 8 | 1559 | 113 | |
More Information
Set Membership: |
|

LOC4339037
Genomic Map