Assay Details
| Gene Symbol: LOC4339144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782728.2 | XP_015638214.1 | 4 - 5 | 1086 | 71 | |
| XM_015782730.2 | XP_015638216.1 | 4 - 5 | 1074 | 71 | ||
| GenBank mRNA | AK073521.1 | - | 5 - 6 | 1110 | 71 | |
More Information
Set Membership: |
|

LOC4339144
Genomic Map