Assay Details
| Gene Symbol: LOC4339144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782728.2 | XP_015638214.1 | 5 - 6 | 1275 | 91 | |
| XM_015782729.2 | XP_015638215.1 | 5 - 6 | 1244 | 91 | ||
| XM_015782730.2 | XP_015638216.1 | 5 - 6 | 1263 | 91 | ||
| GenBank mRNA | AK073521.1 | - | 6 - 7 | 1299 | 91 | |
More Information
Set Membership: |
|

LOC4339144
Genomic Map