Assay Details
| Gene Symbol: LOC4339144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782728.2 | XP_015638214.1 | 7 - 8 | 1581 | 69 | |
| XM_015782729.2 | XP_015638215.1 | 7 - 8 | 1550 | 69 | ||
| XM_015782730.2 | XP_015638216.1 | 7 - 8 | 1476 | 69 | ||
| GenBank mRNA | AK073521.1 | - | 7 - 8 | 1682 | 69 | |
More Information
Set Membership: |
|

LOC4339144
Genomic Map