Assay Details
| Gene Symbol: LOC4339168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784352.2 | XP_015639838.1 | 2 - 3 | 2342 | 148 | |
| XM_015784353.2 | XP_015639839.1 | 2 - 3 | 2345 | 148 | ||
| GenBank mRNA | AK069035.1 | - | 2 - 3 | 2348 | 148 | |
More Information
Set Membership: |
|

LOC4339168
Genomic Map