Assay Details
| Gene Symbol: LOC4339212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785116.2 | XP_015640602.1 | 12 - 13 | 1989 | 75 | |
| GenBank mRNA | AK060635.1 | - | 5 - 6 | 1177 | 75 | |
| AK100622.1 | - | 11 - 12 | 1995 | 75 | ||
More Information
Set Membership: |
|

LOC4339212
Genomic Map