Assay Details
| Gene Symbol: LOC4339216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783880.2 | XP_015639366.1 | 4 - 5 | 528 | 69 | |
| GenBank mRNA | AK058219.1 | - | 4 - 5 | 462 | 69 | |
| AK070301.1 | - | 3 - 4 | 1141 | 69 | ||
| BT128929.1 | - | 4 - 5 | 461 | 69 | ||
| BT129141.1 | - | 4 - 5 | 461 | 69 | ||
| BT129342.1 | - | 4 - 5 | 458 | 69 | ||
| BT129822.1 | - | 4 - 5 | 455 | 69 | ||
| BT130575.1 | - | 4 - 5 | 455 | 69 | ||
| BT131500.1 | - | 4 - 5 | 463 | 69 | ||
More Information
Set Membership: |
|

LOC4339216
Genomic Map