Assay Details
| Gene Symbol: LOC4339223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783658.2 | XP_015639144.1 | 1 - 2 | 508 | 69 | |
| GenBank mRNA | AK059489.1 | - | 1 - 2 | 513 | 69 | |
| AK068896.1 | - | 1 - 2 | 374 | 69 | ||
More Information
Set Membership: |
|

LOC4339223
Genomic Map