Assay Details
| Gene Symbol: LOC4339933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787731.2 | XP_015643217.1 | 3 - 4 | 350 | 131 | |
| GenBank mRNA | AK058985.1 | - | 3 - 4 | 350 | 131 | |
| AK120746.1 | - | 3 - 4 | 320 | 131 | ||
More Information
Set Membership: |
|

LOC4339933
Genomic Map