Assay Details
| Gene Symbol: LOC4339975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788065.2 | XP_015643551.1 | 11 - 12 | 1095 | 115 | |
| XM_015788066.2 | XP_015643552.1 | 11 - 12 | 1095 | 115 | ||
| XM_015788067.2 | XP_015643553.1 | 11 - 12 | 1095 | 115 | ||
| GenBank mRNA | AK122004.1 | - | 11 - 12 | 1043 | 115 | |
More Information
Set Membership: |
|

LOC4339975
Genomic Map