Assay Details
| Gene Symbol: LOC4339975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788065.2 | XP_015643551.1 | 15 - 16 | 1613 | 112 | |
| GenBank mRNA | AK122004.1 | - | 15 - 16 | 1561 | 112 | |
More Information
Set Membership: |
|

LOC4339975
Genomic Map