Assay Details
| Gene Symbol: LOC4340037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785999.2 | XP_015641485.1 | 21 - 22 | 2136 | 102 | |
| GenBank mRNA | AK100230.1 | - | 21 - 22 | 2082 | 102 | |
More Information
Set Membership: |
|

LOC4340037
Genomic Map