Assay Details
| Gene Symbol: LOC4340037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785998.2 | XP_015641484.1 | 3 - 4 | 312 | 64 | |
| XM_015785999.2 | XP_015641485.1 | 3 - 4 | 312 | 64 | ||
| GenBank mRNA | AK100230.1 | - | 3 - 4 | 258 | 64 | |
More Information
Set Membership: |
|

LOC4340037
Genomic Map