Assay Details
| Gene Symbol: LOC4340886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788282.2 | XP_015643768.1 | 12 - 13 | 2914 | 68 | |
| XM_015788283.2 | XP_015643769.1 | 9 - 10 | 2081 | 68 | ||
| GenBank mRNA | AK068880.1 | - | 10 - 11 | 3405 | 68 | |
More Information
Set Membership: |
|

LOC4340886
Genomic Map