Assay Details
| Gene Symbol: LOC4340886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788282.2 | XP_015643768.1 | 1 - 2 | 435 | 97 | |
| GenBank mRNA | AK068880.1 | - | 1 - 2 | 439 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788282.2 transcripts.
Set Membership: |
|

LOC4340886
Genomic Map