Assay Details
| Gene Symbol: LOC4340886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788282.2 | XP_015643768.1 | 5 - 6 | 1067 | 87 | |
| XM_015788283.2 | XP_015643769.1 | 2 - 3 | 234 | 87 | ||
| GenBank mRNA | AK068880.1 | - | 4 - 5 | 1433 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788283.2 transcripts.
Set Membership: |
|

LOC4340886
Genomic Map