Assay Details
| Gene Symbol: LOC4340886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788282.2 | XP_015643768.1 | 11 - 12 | 2839 | 77 | |
| XM_015788283.2 | XP_015643769.1 | 8 - 9 | 2006 | 77 | ||
| GenBank mRNA | AK068880.1 | - | 9 - 10 | 3330 | 77 | |
More Information
Set Membership: |
|

LOC4340886
Genomic Map