Assay Details
| Gene Symbol: LOC4340957 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785558.2 | XP_015641044.1 | 2 - 3 | 299 | 71 | |
| GenBank mRNA | AK059915.1 | - | 1 - 2 | 291 | 71 | |
| U19030.1 | - | 1 - 2 | 346 | 71 | ||
More Information
Set Membership: |
|

LOC4340957
Genomic Map