Assay Details
| Gene Symbol: LOC4340980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787067.2 | XP_015642553.1 | 3 - 4 | 718 | 103 | |
| GenBank mRNA | AK061188.1 | - | 3 - 4 | 724 | 103 | |
| AK067876.1 | - | 3 - 4 | 720 | 103 | ||
| AK099072.1 | - | 3 - 4 | 721 | 103 | ||
More Information
Set Membership: |
|

LOC4340980
Genomic Map