Assay Details
| Gene Symbol: LOC4341018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788787.2 | XP_015644273.1 | 3 - 4 | 544 | 74 | |
| GenBank mRNA | AK063588.1 | - | 3 - 4 | 406 | 74 | |
More Information
Set Membership: |
|

LOC4341018
Genomic Map