Assay Details
| Gene Symbol: LOC4341088 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788372.2 | XP_015643858.1 | 8 - 9 | 2017 | 67 | |
| GenBank mRNA | AK067110.1 | - | 8 - 9 | 1754 | 67 | |
More Information
Set Membership: |
|

LOC4341088
Genomic Map