Assay Details
| Gene Symbol: LOC4341186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788704.2 | XP_015644190.1 | 8 - 9 | 2402 | 90 | |
| GenBank mRNA | AK241349.1 | - | 4 - 5 | 458 | 90 | |
More Information
Set Membership: |
|

LOC4341186
Genomic Map