Assay Details
| Gene Symbol: LOC4337322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781088.2 | XP_015636574.1 | 12 - 13 | 2502 | 122 | |
| GenBank mRNA | AY224540.1 | - | 11 - 12 | 1770 | 122 | |
More Information
Set Membership: |
|

LOC4337322
Genomic Map