Assay Details
| Gene Symbol: LOC4337322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781088.2 | XP_015636574.1 | 15 - 16 | 2941 | 148 | |
| GenBank mRNA | AK073209.1 | - | 13 - 14 | 2139 | 148 | |
| AY224540.1 | - | 14 - 15 | 2209 | 148 | ||
More Information
Set Membership: |
|

LOC4337322
Genomic Map