Assay Details
| Gene Symbol: LOC4337322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781088.2 | XP_015636574.1 | 19 - 20 | 3648 | 117 | |
| GenBank mRNA | AK073001.1 | - | 2 - 3 | 236 | 117 | |
| AK073209.1 | - | 17 - 18 | 2845 | 117 | ||
| AY224540.1 | - | 18 - 19 | 2916 | 117 | ||
More Information
Set Membership: |
|

LOC4337322
Genomic Map