Assay Details
| Gene Symbol: LOC4337322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781088.2 | XP_015636574.1 | 20 - 21 | 3750 | 156 | |
| GenBank mRNA | AK073001.1 | - | 3 - 4 | 338 | 156 | |
| AK073209.1 | - | 18 - 19 | 2947 | 156 | ||
| AY224540.1 | - | 19 - 20 | 3018 | 156 | ||
More Information
Set Membership: |
|

LOC4337322
Genomic Map