Assay Details
| Gene Symbol: LOC4337322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781088.2 | XP_015636574.1 | 22 - 23 | 4218 | 93 | |
| GenBank mRNA | AK073001.1 | - | 5 - 6 | 806 | 93 | |
| AK073209.1 | - | 20 - 21 | 3415 | 93 | ||
| AY224540.1 | - | 21 - 22 | 3486 | 93 | ||
More Information
Set Membership: |
|

LOC4337322
Genomic Map