Assay Details
| Gene Symbol: LOC4337322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781088.2 | XP_015636574.1 | 7 - 8 | 1767 | 104 | |
| GenBank mRNA | AK073209.1 | - | 5 - 6 | 964 | 104 | |
| AY224540.1 | - | 6 - 7 | 1035 | 104 | ||
More Information
Set Membership: |
|

LOC4337322
Genomic Map