Assay Details
| Gene Symbol: LOC4337322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781088.2 | XP_015636574.1 | 10 - 11 | 2142 | 98 | |
| GenBank mRNA | AK073209.1 | - | 8 - 9 | 1340 | 98 | |
| AY224540.1 | - | 9 - 10 | 1410 | 98 | ||
More Information
Set Membership: |
|

LOC4337322
Genomic Map