Assay Details
| Gene Symbol: LOC4338358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783699.2 | XP_015639185.1 | 1 - 2 | 987 | 93 | |
| GenBank mRNA | AK064795.1 | - | 1 - 2 | 422 | 93 | |
More Information
Set Membership: |
|

LOC4338358
Genomic Map