Assay Details
| Gene Symbol: LOC4338358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783699.2 | XP_015639185.1 | 3 - 4 | 1515 | 111 | |
| GenBank mRNA | AK060791.1 | - | 1 - 2 | 251 | 111 | |
| AK064795.1 | - | 3 - 4 | 950 | 111 | ||
More Information
Set Membership: |
|

LOC4338358
Genomic Map