Assay Details
| Gene Symbol: LOC4338358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783699.2 | XP_015639185.1 | 4 - 5 | 1866 | 72 | |
| GenBank mRNA | AK060791.1 | - | 2 - 3 | 600 | 72 | |
| AK064795.1 | - | 4 - 5 | 1301 | 72 | ||
More Information
Set Membership: |
|

LOC4338358
Genomic Map