Assay Details
| Gene Symbol: LOC4338358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783699.2 | XP_015639185.1 | 5 - 6 | 1945 | 64 | |
| GenBank mRNA | AK060791.1 | - | 3 - 4 | 679 | 64 | |
| AK064795.1 | - | 5 - 6 | 1380 | 64 | ||
More Information
Set Membership: |
|

LOC4338358
Genomic Map