Assay Details
| Gene Symbol: LOC4338405 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782645.2 | XP_015638131.1 | 7 - 8 | 1331 | 68 | |
| XM_015782646.2 | XP_015638132.1 | 7 - 8 | 1331 | 68 | ||
| GenBank mRNA | AK100483.1 | - | 6 - 7 | 1396 | 68 | |
More Information
Set Membership: |
|

LOC4338405
Genomic Map