Assay Details
| Gene Symbol: LOC4339633 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783944.2 | XP_015639430.1 | 1 - 2 | 175 | 153 | |
| GenBank mRNA | AK061645.1 | - | 1 - 2 | 181 | 153 | |
| AK099472.1 | - | 1 - 2 | 182 | 153 | ||
| AK100389.1 | - | 1 - 2 | 169 | 153 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015783944.2 transcripts.
Set Membership: |
|

LOC4339633
Genomic Map