Assay Details
| Gene Symbol: LOC4339633 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783944.2 | XP_015639430.1 | 2 - 3 | 587 | 146 | |
| GenBank mRNA | AK061645.1 | - | 2 - 3 | 593 | 146 | |
| AK099472.1 | - | 2 - 3 | 594 | 146 | ||
| AK100389.1 | - | 2 - 3 | 580 | 145 | ||
| AY366480.1 | - | 1 - 2 | 475 | 146 | ||
More Information
Set Membership: |
|

LOC4339633
Genomic Map