Assay Details
| Gene Symbol: LOC4339633 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783944.2 | XP_015639430.1 | 4 - 5 | 781 | 149 | |
| GenBank mRNA | AK061645.1 | - | 4 - 5 | 787 | 149 | |
| AK099472.1 | - | 4 - 5 | 788 | 149 | ||
| AK100389.1 | - | 4 - 5 | 774 | 149 | ||
| AY366480.1 | - | 3 - 4 | 669 | 149 | ||
More Information
Set Membership: |
|

LOC4339633
Genomic Map