Assay Details
| Gene Symbol: LOC4339633 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783944.2 | XP_015639430.1 | 8 - 9 | 1390 | 85 | |
| GenBank mRNA | AK061645.1 | - | 8 - 9 | 1396 | 85 | |
| AK099472.1 | - | 8 - 9 | 1397 | 85 | ||
| AK100389.1 | - | 8 - 9 | 1384 | 85 | ||
| AY366480.1 | - | 7 - 8 | 1278 | 85 | ||
More Information
Set Membership: |
|

LOC4339633
Genomic Map