Assay Details
| Gene Symbol: LOC4339633 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783944.2 | XP_015639430.1 | 9 - 10 | 1464 | 62 | |
| GenBank mRNA | AK061645.1 | - | 9 - 10 | 1470 | 62 | |
| AK099472.1 | - | 9 - 10 | 1471 | 62 | ||
| AK100389.1 | - | 9 - 10 | 1458 | 62 | ||
| AY366480.1 | - | 8 - 9 | 1352 | 62 | ||
More Information
Set Membership: |
|

LOC4339633
Genomic Map