Assay Details
Gene Symbol: LOC4339708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785272.2 | XP_015640758.1 | 3 - 4 | 984 | 78 | |
GenBank mRNA | AK108477.1 | - | 1 - 2 | 101 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
