Assay Details
| Gene Symbol: LOC4340478 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787838.2 | XP_015643324.2 | 10 - 11 | 1628 | 67 | |
| XM_015787840.2 | XP_015643326.1 | 6 - 7 | 1031 | 67 | ||
| XM_015787841.2 | XP_015643327.1 | 5 - 6 | 924 | 67 | ||
| GenBank mRNA | AK064752.1 | - | 10 - 11 | 1566 | 67 | |
More Information
Set Membership: |
|

LOC4340478
Genomic Map