Assay Details
| Gene Symbol: LOC4340478 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787838.2 | XP_015643324.2 | 9 - 10 | 1524 | 69 | |
| XM_015787840.2 | XP_015643326.1 | 5 - 6 | 927 | 69 | ||
| XM_015787841.2 | XP_015643327.1 | 4 - 5 | 820 | 69 | ||
| GenBank mRNA | AK064752.1 | - | 9 - 10 | 1462 | 69 | |
More Information
Set Membership: |
|

LOC4340478
Genomic Map