Assay Details
| Gene Symbol: LOC4341462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788395.1 | XP_015643881.1 | 2 - 3 | 1002 | 64 | |
| XM_015788396.1 | XP_015643882.1 | 2 - 3 | 1002 | 64 | ||
| GenBank mRNA | AK068856.1 | - | 2 - 3 | 970 | 64 | |
| AK104955.1 | - | 2 - 3 | 962 | 64 | ||
| EU781632.1 | - | 2 - 3 | 670 | 64 | ||
More Information
Set Membership: |
|

LOC4341462
Genomic Map