Assay Details
| Gene Symbol: LOC4341462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788395.1 | XP_015643881.1 | 3 - 4 | 1106 | 72 | |
| GenBank mRNA | AK068856.1 | - | 3 - 4 | 1074 | 72 | |
| AK104955.1 | - | 3 - 4 | 1066 | 72 | ||
| EU781632.1 | - | 3 - 4 | 774 | 72 | ||
More Information
Set Membership: |
|

LOC4341462
Genomic Map