Assay Details
| Gene Symbol: LOC4341464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785567.2 | XP_015641053.1 | 1 - 2 | 895 | 93 | |
| XM_015785570.2 | XP_015641056.1 | 1 - 2 | 895 | 93 | ||
| XM_015785573.2 | XP_015641059.1 | 1 - 2 | 895 | 93 | ||
| GenBank mRNA | AK106905.1 | - | 2 - 3 | 520 | 93 | |
More Information
Set Membership: |
|

LOC4341464
Genomic Map