Assay Details
| Gene Symbol: LOC4341477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787598.1 | XP_015643084.1 | 10 - 11 | 1115 | 122 | |
| GenBank mRNA | EU703803.1 | - | 10 - 11 | 1016 | 122 | |
| EU703804.1 | - | 9 - 10 | 971 | 122 | ||
More Information
Set Membership: |
|

LOC4341477
Genomic Map