Assay Details
| Gene Symbol: LOC4341477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787598.1 | XP_015643084.1 | 6 - 7 | 656 | 85 | |
| GenBank mRNA | AK102270.1 | - | 6 - 7 | 656 | 85 | |
| EU703803.1 | - | 6 - 7 | 557 | 85 | ||
| EU703804.1 | - | 5 - 6 | 512 | 85 | ||
More Information
Set Membership: |
|

LOC4341477
Genomic Map