Assay Details
| Gene Symbol: LOC4341727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789024.2 | XP_015644510.1 | 4 - 5 | 750 | 116 | |
| XM_015789025.2 | XP_015644511.1 | 4 - 5 | 750 | 116 | ||
| GenBank mRNA | AK243323.1 | - | 4 - 5 | 663 | 116 | |
More Information
Set Membership: |
|

LOC4341727
Genomic Map